Learn More
Fisherbrand™ Catalyst Tygon E-Food Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15581345
  • £194.39 / Pack of 12
Estimated Shipment
13-07-2021
Add to basket
Description
Description
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Specifications
Stopped tubing assembly
6.4 mm
2.4 mm