Learn More
Fisherbrand™ Catalyst Tygon E-Food Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™
Additional Details : Weight : 0.16200kg
 View more versions of this product
Product Code. 15501375
  • £115.00 / Pack of 12
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • Time-saving assembly
  • Four formulations for range of chemical compatibility
  • Use with Catalyst extension coils/tubing
Specifications
Specifications
Stopped Tubing Assembly
3.2 mm