Learn More
Fisherbrand™ Catalyst PharMed™ BPT Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand: Fisherbrand™
Additional Details : Weight : 0.04500kg
Description
- Available in Platinum-Cured Silicone, TPEs, and PVC
- Ensures optimal amount of tubing is in pump head
Specifications
Stopped Tubing Assembly | |
3.2 mm | |
1.6 mm |