Learn More
Fisherbrand™ Catalyst PharMed™ BPT Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15501335
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Specifications
Stopped Tubing Assembly
9.5 mm
2.4 mm