Learn More
Fisherbrand™ Catalyst PharMed™ BPT Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15561325
  • £270.14 / Pack of 12
Estimated Shipment
15-12-2020
Add to basket
Description
Description
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Specifications
Stopped Tubing Assembly
8 mm
1.6 mm