missing translation for 'onlineSavingsMsg'
Learn More

Castanospermine, Tocris Bioscience™

Glucosidases α and β inhibitor

Brand:  Tocris (Bio-Techne) 075950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13487963

  • £336.61 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Potent inhibitor of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids). Inhibits HIV syncytium formation and replication.
Specifications

Chemical Identifiers

79831-76-8
189.211
[1S-(1α,6β,7α,8β,8aβ)]-Octahydro-1,6,7,8-indolizinetetrol
CHEBI:27860
C1CN2CC(C(C(C2C1O)O)O)O
C8H15NO4
JDVVGAQPNNXQDW-TVNFTVLESA-N
54445
(1S,6S,7R,8R,8aR)-1,2,3,5,6,7,8,8a-octahydroindolizine-1,6,7,8-tetrol

Specifications

Castanospermine
79831-76-8
C8H15NO4
JDVVGAQPNNXQDW-TVNFTVLESA-N
(1S,6S,7R,8R,8aR)-1,2,3,5,6,7,8,8a-octahydroindolizine-1,6,7,8-tetrol
54445
189.21
50mg
>98%
[1S-(1α,6β,7α,8β,8aβ)]-Octahydro-1,6,7,8-indolizinetetrol
C1CN2CC(C(C(C2C1O)O)O)O
189.211
CHEBI:27860
Videos