missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-2 Substrate, chromogenic, Thermo Scientific™
For cellular and molecular biology applications
£154.00 - £256.00
Specifications
Color | White to off-white |
---|---|
Form | Lyophilized |
Substrate Type | Caspase Substrate |
Molecular Weight (g/mol) | 765.72 |
Storage Requirements | -30° to -10°C |
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
15468559
|
Thermo Scientific Alfa Aesar
J66974.MA |
10mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15478559
|
Thermo Scientific Alfa Aesar
J66974.MB |
25mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White to off-white | |
Caspase Substrate | |
-30° to -10°C |
Lyophilized | |
765.72 |
For Research Use Only