Learn More
Caspase 14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555497
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RLALILCVTK AREGSEEDLD ALEHMFRQLR FESTMKRDPT AEQFQEELEK FQQAIDSRED PVSCAFVVLM AHGREGFLKG EDGEMVKLEN LFEALNNKNC QALRAKPKVY IIQACRGEQR DPGETVGGDE IVM Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
Caspase 14 Polyclonal antibody specifically detects Caspase 14 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Caspase 14 | |
Polyclonal | |
Unconjugated | |
Casp14 | |
apoptosis-related cysteine protease; CASP14; CASP-14; caspase 14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase14; caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form; caspase-14; LOW QUALITY PROTEIN: caspase-14; MICE; Mini-ICE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23581 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P31944 | |
Casp14 | |
Recombinant protein corresponding to Human Caspase 14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only