Learn More
Alfa Aesar™ Casein Sodium Salt
For research use
Brand:  Alfa Aesar™ J65590.A7
Code : NEW
Additional Details : CAS Number : 9005-46-3 Weight : 5.00000kg
 View more versions of this product
Product Code. 15496979
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only
Specifications
Specifications
Casein is frequently used as a blocking agent in immunochemistry experiments. It is also used in the recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
Lyophilized powder
Bovine milk
Casein sodium salt
Lyophilized Powder
5kg
RUO
No
Videos