Alfa Aesar™ Casein Sodium Salt

For research use

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J65590.A1

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9005-46-3 Weight: 1.00000kg



Disclaimers: For Research Use Only

Product Code. 15486979

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Product Type Casein sodium salt
Format Lyophilized powder
Regulatory Status RUO
Formulation Lyophilized powder
Source Bovine milk
Quantity 1kg
Recombinant No
For Use With (Application) Casein is frequently used as a blocking agent in immunochemistry experiments. It is also used in the recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
Alternative Products