missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Casein, High Nitrogen, 95%

For research use

Brand:  Alfa Aesar™ J64482.36

Code : NEW

Additional Details : CAS Number : 9000-71-9 Weight : 0.50000kg

 View more versions of this product

Product Code. 15486409

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Lyophilized Powder
95%
Bovine milk
Casein, high nitrogen, 95%
Lyophilized powder
500g
RUO
No
Videos
SDS

For Research Use Only