Alfa Aesar™ Casein, Hammarsten grade


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J64214.36

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9000-71-9 Weight: 0.50000kg



Product Code. 15446269

Quantity Price
1 £ 54.480000000000004 / 500g
Estimated Shipment date
from Supplier 16-05-2017
Add to basket


Description and Specification

Specification

Color Off-white to Pale Yellow
Format Powder
Product Type Blocking Buffer
Quantity 500g
Sensitivity Hygroscopic
CAS 9000-71-9
Synonym Casein, Hammerstein Grade
Odor Odorless
Documentation

Technical documents