missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Casein Blocking Buffer, 3% in TBS, with 0.02% sodium azide

For Western blots and ELISA

Brand:  Alfa Aesar™ J62935.AK

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15485229

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

For Western blots and ELISA
Blocking Buffer
−30° to −10°C
Liquid
250mL
Videos
SDS