Alfa Aesar™ Casein Blocking Buffer, 3% in PBS, with 0.02% sodium azide

For Western blots and ELISA

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J63453.AK

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg



Product Code. 15415699

Quantity Price
1 £ 31.06 / 250mL
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Specification

Product Type Blocking Buffer
Storage Requirements −30° to −10°C
Format Liquid
Quantity 250mL
Applications For Western blots and ELISA