missing translation for 'onlineSavingsMsg'
Learn More

(-)-Caryophyllene oxide, 95%, ACROS Organics™

Brand:  Acros Organics 325082500

Code : 29

Additional Details : CAS Number : 1139-30-6 Weight : 0.25000kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10636184

  • £146.30 / 250g


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

(-)-Caryophyllene oxide
94.0
≥94%
C15H24O
MFCD00134216
1139-30-6
100.0
95%
220.35
(1R,4R,6R,10S)-4,12,12-Trimethyl-9-methylene-5-oxatricyclo[8.2.0.0(4,6)]dodecane

Specifications

(-)-Caryophyllene oxide
White
250g
94.0
96%g/mL
Crystalline Powder
95%
C15H24O
MFCD00134216
-40° to -70° (20°C, 589nm) (c=10, EtOH)
−55° (20°C 589nm,c=10,EtOH)
55° to 62°C
1139-30-6
100.0
Authentic
≥94%
Glass bottle
220.35
17, IV, 392
(1R,4R,6R,10S)-4,12,12-Trimethyl-9-methylene-5-oxatricyclo[8.2.0.0(4,6)]dodecane
Videos
SDS