missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15360974
|
RETSCH
02.225.0088 |
For 50 mL Agate Grinding Set |
£131.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15380974
|
RETSCH
02.225.0090 |
For 100 mL Tungsten Carbide Grinding Set |
£131.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15350974
|
RETSCH
02.225.0087 |
For RS 200 and 50 mL Hardened Steel, Tungsten Carbide, Steel Grinding Set |
£131.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Specifications
Carry Handle Insert |