missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Carpenter 20™ gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire

Brand:  Alfa Aesar™ 046339.FL

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 75 x 75mm

 View more versions of this product

Product Code. 15427185

  • £13.10 / Each


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Carpenter 20™ gauze
40 mesh woven from 0.25mm (0.01 in.) dia. wire

Specifications

Carpenter 20™ gauze
75 x 75mm
Odorless
40 Mesh
40 mesh woven from 0.25mm (0.01 in.) dia. wire
Wire Cloth
Videos
SDS