missing translation for 'onlineSavingsMsg'
Learn More

Carpenter 20™ gauze, 24 mesh woven from 0.36mm (0.014 in.) dia. wire, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 046740.HB

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15497295

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Carpenter 20™ gauze
300 x 300 mm
Odorless
24 mesh woven from 0.36mm (0.014 in.) dia. wire
24 Mesh
Wire Cloth
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers