missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken Caronte Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Caronte |
---|---|
Clone | 156808 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Caronte Monoclonal specifically detects Caronte in Chicken samples. It is validated for Western Blot.Specifications
Caronte | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Chicken | |
NP_990154 | |
9350 | |
Mouse myeloma cell line NS0-derived recombinant chicken Caronte Asp16-Asn272 Accession # NP_990154 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects chicken Caronte in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Cerberus, rhDAN, or recombinant mouse Gremlin is observed. |
156808 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Chicken Caronte | |
CER1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only