missing translation for 'onlineSavingsMsg'
Learn More

Carnauba wax, refined, No. 1, yellow, ACROS Organics™

Brand:  Acros Organics 222380050

Code : NEW

Additional Details : CAS Number : 8015-86-9 Weight : 5.00000kg

Packaging Plastic drum
Quantity 5kg

 View more versions of this product

Product Code. 13468478

  • £219.90 / 5kg

Estimated Shipment date
from Supplier 04-03-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

8015-86-9
MFCD00130724
Brazil wax

Specifications

2 to 7 (mg KOH/g)
282°C
Plastic drum
78 to 95mg KOH/g
81.0°C to 86.0°C
8015-86-9
15,1846
Solubility in water: insoluble. Other solubilities: soluble in boiling alcohol,diethyl ether,benzene,,toluene,chloroform,ethyl acetate,turpentine,,petroleum ether,carbon disulfide;,soluble in alkalies
Carnauba wax
0.25% max.
0.25% max
Yellow
5kg
MFCD00130724
Brazil wax
Powder or Flakes
Videos
Documents