Learn More
Tocris Bioscience™ Carmoxirole hydrochloride
Selective, peripherally acting D2-like agonist
Brand:  Tocris Bioscience™ 219310
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13440764
  • £127.89 / 10mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Selective, peripherally acting dopamine D2 receptor agonist. Modulates noradrenalin release and sympathetic activation. Displays antihypertensive properties in vivo.
Specifications
Specifications
Carmoxirole hydrochloride
>98
10mg
Carmoxirole hydrochloride
410.94