Learn More
Carl Zeiss™ Microscope Stage Micrometers
Finely divided stage micrometer for calibrating optical systems with eyepiece graticule patterns. Carl Zeiss™ Microscope Stage Micrometer includes 1mm and 1/100mm intervals.
POA
Specifications
Type | Stage Micrometer |
---|---|
Includes | Stage micrometer, 5 lines with 1mm intervals in y-direction |
Description
- For calibrating optical systems with eyepiece graticule patterns
- 5 lines with 1mm intervals in y-direction
- 100 lines with 1/100mm intervals in y-direction
Specifications
Stage Micrometer | |
Stage micrometer, 5 lines with 1mm intervals in y-direction |