Learn More
CARKD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA552705
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PNAVHEVEKW LPRLHALVVG PGLGRDDALL RNVQGILEVS KARDIPVVID ADGLWLVAQQ PALIHGYRKA VLTPNHVEFS RLYDAVLRGP MDSDDSHGSV LRLSQALGNV TVVQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.
CARKD Polyclonal antibody specifically detects CARKD in Human samples. It is validated for Immunohistochemistry.Specifications
CARKD | |
Polyclonal | |
Unconjugated | |
NAXD | |
0710008K08Rik; 2810407E01Rik; ATP-dependent (S)-NAD(P)H-hydrate dehydratase; ATP-dependent (S)-NAD(P)H-hydrate dehydratase {ECO:0000255; ATP-dependent NAD(P)H-hydrate dehydratase; ATP-dependent NAD(P)HX dehydratase; ATP-dependent NAD(P)HX dehydratase {ECO:0000255; carbohydrate kinase domain containing; Carbohydrate kinase domain-containing protein; carbohydrate kinase domain-containing protein {ECO:0000255; CARKD; HAMAP-Rule:MF_03157}; LP3298; NAD(P)HX dehydratase; Naxd; RGD1562691; zgc:171429 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55739 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IW45 | |
NAXD | |
Recombinant protein corresponding to Human CARKD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only