missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550320
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total CARF / ALS2CR8 protein.
CARF Polyclonal antibody specifically detects CARF in Human samples. It is validated for Western Blot.Specifications
CARF | |
Polyclonal | |
Unconjugated | |
CARF | |
Als2cr8; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 8; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein homolog; calcium response factor; calcium responsive transcription factor; calcium-response factor; calcium-response factor CaRF; calcium-responsive transcription factor; CaRF; ECBRC-FC1; ECBRC-FC2; NYD-SP24; testis development protein NYD-SP24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79800 | |
-20°C | |
Liquid |
Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N187 | |
CARF | |
Fusion protein corresponding to residues near the C terminal of human calcium responsive transcription factor | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |