Learn More
Ratiolab™ Cardboard Cryo Boxes With Waterproof Plastic Coating
Combine these boxes with different grid inserts for ultra-low temperature storage. Ratiolab™ Cardboard Cryo Boxes With Waterproof Plastic Coating are ideal for sample storage and transportation.
£2.81 - £109.00
Specifications
Material | Cardboard |
---|---|
Type | Cryo Box |
For Use With (Application) | Tubes from dia. 6mm to dia. 28.5mm |
Quantity | 10 Pack |
Product Code | Brand | Color | Length (Metric) | Width (Metric) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Length (Metric) | Width (Metric) | Height (Metric) | Price | Quantity & Availability | ||
10212343
|
Ratiolab™
5142002 |
Blue | 136 mm | 136 mm | 100 mm |
£70.35
Pack of 10 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
10553575
|
Ratiolab™
5142003 |
Red | 136 mm | 136 mm | 100 mm |
£71.10
Pack of 10 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||
10434363
|
Ratiolab™
5142004 |
Green | 136 mm | 136 mm | 100 mm |
£72.10
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10632254
|
Ratiolab™
5142001 |
White | 136 mm | 136 mm | 100 mm |
£53.85
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10232483
|
Ratiolab™
5142005 |
Yellow | 136 mm | 136 mm | 100 mm |
£72.10
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
12625785
|
Ratiolab™
5132132 |
Blue | 133 mm | 133 mm | 130 mm |
£109.00
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10707123
|
Ratiolab™
5142132 |
Blue | 136 mm | 136 mm | 130 mm |
£58.90
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10115974
|
Ratiolab™
5132131 |
White | 133 mm | 133 mm | 130 mm |
£95.85
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10501085
|
Ratiolab™
5142131 |
White | 136 mm | 136 mm | 130 mm |
£58.90
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10304633
|
Ratiolab™
5142135 |
Yellow | 136 mm | 136 mm | 130 mm |
£58.90
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10150803
|
Ratiolab™
5142321 |
White | 136 mm | 136 mm | 32 mm |
£42.15
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
15293979
|
Ratiolab™
5132322 |
Blue | 133 mm | 133 mm | 32 mm |
£54.40
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10799054
|
Ratiolab™
5142325 |
Yellow | 136 mm | 136 mm | 32 mm |
£51.85
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
11893892
|
Ratiolab™
5142502 |
Blue | 136 mm | 136 mm | 50 mm |
£4.23
Each Qty Price Break Available |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
10222673
|
Ratiolab™
5142502 |
Blue | 136 mm | 136 mm | 50 mm |
£45.05
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10691674
|
Ratiolab™
5142504 |
Green | 136 mm | 136 mm | 50 mm |
£45.05
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10141093
|
Ratiolab™
5142503 |
Red | 136 mm | 136 mm | 50 mm |
£45.05
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
15210568
|
Ratiolab™
5132501 |
White | 133 mm | 133 mm | 50 mm |
£29.20
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10797453
|
Ratiolab™
5142501 |
White | 136 mm | 136 mm | 50 mm |
£32.80
Pack of 10 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||
15971641
|
Ratiolab™
5132505 |
Yellow | 133 mm | 133 mm | 50 mm |
£46.30
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10406283
|
Ratiolab™
5142505 |
Yellow | 136 mm | 136 mm | 50 mm |
£45.05
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
10707293
|
Ratiolab™
5142754 |
Green | 136 mm | 136 mm | 75 mm |
£66.80
Pack of 10 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||
10141043
|
Ratiolab™
5142751 |
White | 136 mm | 136 mm | 75 mm |
£34.05
Pack of 10 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||
10400395
|
Ratiolab™
5132501 |
White | 133 mm | 133 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11883892
|
Ratiolab™
5142001 |
White | 136 mm | 136 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11803902
|
Ratiolab™
5142503 |
Red | 136 mm | 136 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Ultra low temperature-resistant cardboard
- For sample storage
- Temperature-resistant down to -80°C
- Water-repellent finish
- Packaging in practical folding boxes
Specifications
Cardboard | |
Cryo Box | |
Tubes from dia. 6mm to dia. 28.5mm | |
10 Pack |