missing translation for 'onlineSavingsMsg'
Learn More

Human Carboxypeptidase X1/CPXM1 Antibody, R&D Systems™

Goat Polyclonal Antibody

POA

Specifications

Antigen Carboxypeptidase X1/CPXM1
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
View More Specs

Description

Description

Carboxypeptidase X1/CPXM1 Polyclonal specifically detects Carboxypeptidase X1/CPXM1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Carboxypeptidase X1/CPXM1
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Polyclonal
Goat
Human
carboxypeptidase X (M14 family), carboxypeptidase X (M14 family), member 1, CPX1, CPX-1, CPX1EC 3.4.17.-, CPXM, CPXM1, EC 3.4.17, EC 3.4.17.10, EC 3.4.17.3, Metallocarboxypeptidase CPX-1, probable carboxypeptidase X1
CPXM1
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot, Immunoprecipitation
Unconjugated
RUO
Q96SM3
56265
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase X1/CPXM1 Ala21-Asp734 Accession # Q96SM3
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects human Carboxypeptidase X1/CPXM1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse CPXM is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only