missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase D Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA562688

Additional Details : Weight : 0.02000kg

Product Code. 15720482

  • £340.00 / 100µL

Estimated Shipment date
from Supplier 18-06-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The metallocarboxypeptidase family of enzymes is divided into 2 subfamilies based on sequence similarities. The pancreatic carboxypeptidase-like and the regulatory B-type carboxypeptidase subfamilies. Carboxypeptidase D has been identified as a regulatory B-type carboxypeptidase. CPD is a homolog of duck gp180, a hepatitis B virus-binding protein. Transcript variants utilizing alternative polyadenylation signals exist for this gene.
Specifications

Specifications

Carboxypeptidase D
Polyclonal
Unconjugated
CPD
Liquid
CPD
Recombit protein corresponding to Human Carboxypeptidase D
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75976
Carboxypeptidase D; Metallocarboxypeptidase D; gp180; CPD
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1362
Videos
Documents

Research Use Only