missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase B1/CPB1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Carboxypeptidase B1/CPB1 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase B1/CPB1 His16-Tyr417 Accession # P15086 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
Carboxypeptidase B1/CPB1 Polyclonal specifically detects Carboxypeptidase B1/CPB1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase B1/CPB1 | |
Goat | |
Polyclonal | |
IgG | |
carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, CPB1, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunoprecipitation | |
RUO |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase B1/CPB1 His16-Tyr417 Accession # P15086 | |
Primary | |
Human | |
Unconjugated | |
1360 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only