missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A2/CPA2 Mouse anti-Human, Clone: 384023, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Carboxypeptidase A2/CPA2 |
---|---|
Clone | 384023 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase A2/CPA2 Leu18-His417 Accession # P48052 |
Host Species | Mouse |
Primary or Secondary | Primary |
Description
Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase A2/CPA2 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase A2/CPA2 Leu18-His417 Accession # P48052 | |
Primary | |
Human | |
Unconjugated | |
1358 | |
Purified | |
Western Blot, Immunoprecipitation | |
RUO |
384023 | |
Mouse | |
Monoclonal | |
IgG2b | |
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), CPA2, EC 3.4.17, EC 3.4.17.15 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only