missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1/CPA1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
POA
Specifications
Antigen | Carboxypeptidase A1/CPA1 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase A1/CPA1 Asn17-Tyr419 Accession # Q7TPZ8 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
Carboxypeptidase A1/CPA1 Polyclonal specifically detects Carboxypeptidase A1/CPA1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Carboxypeptidase A1/CPA1 | |
Goat | |
Polyclonal | |
IgG | |
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, CPA1, EC 3.4.17, EC 3.4.17.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunoprecipitation, Neutralization | |
RUO |
Mouse myeloma cell line NS0-derived recombinant mouse Carboxypeptidase A1/CPA1 Asn17-Tyr419 Accession # Q7TPZ8 | |
Primary | |
Mouse | |
Unconjugated | |
1357 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only