missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carboxymethylecellulose, sodium salt, average M.W. 700000 (DS=0.9)
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Beige |
pH | 6.5 to 8 (1% soln.) |
Physical Form | Crystalline Powder |
Merck Index | 15, 1830 |
Quantity | 100 g |
Percent Purity | ≥99.5% |
Infrared Spectrum | Authentic |
Loss on Drying | 10% max. (as packed) (3 to 5 g, 105°C, 2 hrs) |
Packaging | Glass bottle |
Viscosity | 2500 to 4500 mPa.s (Brookfield, 1% soln., at 25°C) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'