missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethoxylamine HemiHydroChloride, 98%, Thermo Scientific Chemicals

Catalog Number p-3735813
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2921-14-4
C4H11ClN2O6
218.59
MFCD00012955
AXIKLJPWEGNZIG-UHFFFAOYSA-N
o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride
2723609
2-aminooxyacetic acid;hydrochloride
[Cl-].NOCC(O)=O.[NH3+]OCC(O)=O
This item is not returnable. View return policy
2921-14-4
C4H11ClN2O6
218.59
MFCD00012955
AXIKLJPWEGNZIG-UHFFFAOYSA-N
o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride
2723609
2-aminooxyacetic acid;hydrochloride
[Cl-].NOCC(O)=O.[NH3+]OCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (H2NOCH2CO2H)2·HCl
Quantity 2.5 g
Solubility Information Solubility in water: decomposes.
Formula Weight 109.3
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Carboxymethoxylamine hemihydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'