Learn More
Tocris Bioscience™ Carboxy-PTIO, potassium salt
Brand:  Tocris Bioscience™ 0772/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13457983
  • £128.25 / 10mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
A stable, water-soluble free radical that reacts stoichiometrically with NO. Almost twice as strong an inhibitor as N(ω)-nitroarginine or N-methylarginine.
Specifications
Specifications
2-(4-Carboxyphenyl)-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide, potassium salt
C14H16KN2O4
10mg
148819-94-7
Observed MW: 315.38
Soluble to 25 mM in ethanol and to 10 mM in water