missing translation for 'onlineSavingsMsg'
Learn More

Carbonyldihydridotris(triphenylphosphine)ruthenium(II), White crystalline, 99%, Thermo Scientific Chemicals

Catalog Number p-4781843
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
25360-32-1
C55H47OP3Ru
917.97
MFCD00015870 MFCD00015870
MLIYPCQSOXNTLJ-UHFFFAOYSA-N
53384314
carbon monoxide;hydride;ruthenium(2+);triphenylphosphane
[RuH2].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
25360-32-1
C55H47OP3Ru
917.97
MFCD00015870 MFCD00015870
MLIYPCQSOXNTLJ-UHFFFAOYSA-N
53384314
carbon monoxide;hydride;ruthenium(2+);triphenylphosphane
[RuH2].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Carbonyldihydridotris(triphenylphosphine)ruthenium(II) is used as Catalyst for conversion of 1,4-alkynediols into pyrroles, Catalyst for oxidation of primary alcohols to methyl esters, Catalyst for rearrangement of oximes to amides, Catalyst for C-C coupling via directed C-H activation, Catalyst for enyne cyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 161°C to 163°C
Color White
Assay Percent Range 99%
Quantity 0.5 g
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 917.97
Physical Form Crystals
Chemical Name or Material Carbonyldihydridotris(triphenylphosphine)ruthenium(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'