Learn More
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II), Thermo Scientific Chemicals
Description
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) is used as a raw material to synthesize other ru-hydride catalysts. It is involved in the hydrogenation of ring-opening metathesis polymers such as norbornene and other cyclic olefin ROMP polymers used as plastics, resins and optical materials. It can act as an alternative catalyst to rhodium hydrogenation catalyst. It also serves as a catalyst in the hydrogenation of conjugated diene copolymers like acrylonitrile-butadiene rubber (NBR).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 204°C to 206°C (decomposition) |
Color | Gray |
Linear Formula | [(C6H5)3P]3Ru(CO)(Cl)H |
Quantity | 1 g |
Sensitivity | Light sensitive; Very Hygroscopic |
Solubility Information | Insoluble in water,ethanol,and DMSO. Sparingly soluble in benzene,chloroform and acetone. |
Formula Weight | 952.41 |
Physical Form | Crystalline |
Chemical Name or Material | Carbonylchlorohydridotris (triphenylphosphine)ruthenium (II) |
RUO – Research Use Only