missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Brand:  Alfa Aesar™ 044955.14

Code : NEW

Additional Details : CAS Number : 16971-33-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15416645

  • £995.90 / 25g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

Carbonylchlorohydridotris (triphenylphosphine)ruthenium (II)
C55H46ClOP3Ru
952.41
16971-33-8
[(C6H5)3P]3Ru(CO)(Cl)H
MFCD00049804

Specifications

Carbonylchlorohydridotris (triphenylphosphine)ruthenium (II)
25g
Crystalline
[(C6H5)3P]3Ru(CO)(Cl)H
MFCD00049804
204° to 206°C (dec.)
Moisture & Light Sensitive
16971-33-8
C55H46ClOP3Ru
952.41
Gray
Videos
SDS