Alfa Aesar™ Carbonylchlorobis(triphenylphosphine)rhodium(I), Rh 14.9% min


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 039822.06

CARBONYLCHLOROBIS(TRIPHE- NYLPHOSPHINE)RH(I) 5G

UNSPSC: 12141738

Code: NEW

Additional Details:
Additional Details: CAS Number: 13938-94-8 Weight: 0.01000kg


Quantity 5g

Product Code. 11359137

Quantity Price
1 £ 613.0 / 5g
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Carbonylchlorobis(triphenylphosphine)rhodium(I)
CAS 13938-94-8
Linear Formula RhCl(CO)[P(C6H5)3]2
Formula Weight 690.94
MDL Number MFCD00044273
Beilstein 4618896
Synonym Carbonylbis(triphenylphosphine)rhodium(I) chloride

Specification

Formula Weight 690.94
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Carbonylchlorobis(triphenylphosphine)rhodium(I)
Odor Odorless
Physical Form Crystalline
Linear Formula RhCl(CO)[P(C6H5)3]2
MDL Number MFCD00044273
CAS 13938-94-8
Beilstein 4618896
Synonym Carbonylbis(triphenylphosphine)rhodium(I) chloride
Solubility Information Soluble in acetone, chloroform, ethanol
Health Hazard 1 Warning
Color Yellow
Melting Point 224° to 227°C (dec.)
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Concentration Rh 14.9% min.
Quantity 5g
Safety and Handling

Warning

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.