missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I), Rh 20%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
0.25 g
1 g
5 g
Descrizione
Hydroformylation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Color | Yellow |
Quantity | 5 g |
Merck Index | 14,356 |
Solubility Information | Soluble in acetone and chlorinated solvents |
Formula Weight | 492.32 |
Concentration or Composition (by Analyte or Components) | Rh 21% |
Physical Form | Crystals |
Chemical Name or Material | Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'