missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonate solution, 0.22M Carb./ 0.28M Bicarb. for Ion Chromatography, 100mg/L, H2O matrix, Fisher Chemical™
Click to view available options
Quantity:
500 mL
Packaging:
Bottle
Specifications
Concentration | 100mg/L |
Packaging | Bottle |
Quantity | 500 mL |
Type | Single-component Standard Solution |
Solution Contains | 0.22M Carbonate/0.28M Bicarbonate |
Solvent or Matrix | H2O |
Physical Form | Liquid |
Chemical Name or Material | Carbonate solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'