missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Carbon powder, activated, Norit™ D10

Brand:  Alfa Aesar™ H36691.36

Code : NEW

Additional Details : CAS Number : 7440-44-0 Weight : 0.50000kg Transport : UN number : 1362 Chem class : 1 Pack group : III

Quantity 500g

 View more versions of this product

Product Code. 15437687

  • £44.80 / 500g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Description

Description

Applications
Carbon powder is used as such in purifying metals, in electrodes, electrical devices and steel.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

Carbon powder
Activated, Norit™ D10
UN1362
7440-44-0
MFCD00133992

Specifications

Carbon powder
500g
7440-44-0
MFCD00133992
Black
Activated, Norit™ D10
Powder
UN1362
Videos
SDS