missing translation for 'onlineSavingsMsg'
Learn More

Carbon felt, 6.35mm (0.25in) thick, 99.0%, Thermo Scientific Chemicals

Catalog Number p-4781842
Click to view available options
Quantity:
10 x 10 cm
30 x 30 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1333-86-4
C
12.01
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1333-86-4
C
12.01
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N

Typical thermal conductivity of 1.75 BTU in/hr/ft²/°F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 2.26g/cm3
Assay Percent Range 99.0%
Quantity 10 x 10 cm
Chemical Name or Material Carbon

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'