missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Carbon felt, 2.54cm (1.0 in.) thick, 99.0%

Brand:  Alfa Aesar™ 043201.RR

Code : NEW

Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg

Quantity 10 x 10cm

 View more versions of this product

Product Code. 11319268

  • £36.20 / Each


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

Carbon felt
2.54cm (1.0 in.) thick
MFCD00133992

Specifications

Carbon felt
10 x 10cm
99.0%
≈19.1g/10x10cm
2.54cm (1.0 in.) thick
MFCD00133992
Videos
SDS