missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Carbon felt, 1.27cm (0.5 in.) thick, 95%

Brand:  Alfa Aesar™ 043026.RF

Code : NEW

Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg

Quantity 30 x 30cm

 View more versions of this product

Product Code. 11394668

  • £57.10 / Each


Estimated Shipment date
from Supplier 18-01-2018
Add to basket

Specifications

Chemical Identifiers

Carbon felt
1.27cm (0.5 in.) thick
MFCD00133992

Specifications

Carbon felt
30 x 30cm
95%
≈10.6g/10x10cm
1.27cm (0.5 in.) thick
MFCD00133992
Videos
SDS