Learn More
Thermo Scientific Alfa Aesar Carbon felt, 1.27cm (0.5 in.) thick, 95%
Brand: Thermo Scientific Alfa Aesar 043026.RR
Carbon felt, 1.27cm (0.5 in.) thick, 95%
Code : NEW
Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg
Brand: Thermo Scientific Alfa Aesar 043026.RR
Carbon felt, 1.27cm (0.5 in.) thick, 95%
Code : NEW
Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg