Learn More
Carbon disulfide, low benzene, 99.9%, Thermo Scientific Chemicals
Description
Carbon disulfide is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reactive agent for the production of rubber vulcanization accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 112°C |
Assay Percent Range | 99.9% |
Solubility Information | Soluble in alcohol,ether,benzene,oil,chloroform and carbon tetrachloride. |
Formula Weight | 76.14 |
Density | 1.2632 g/mL |
Boiling Point | 46°C |
Quantity | 100 mL |
Flash Point | -30°C (-22°F) |
Physical Form | Liquid |
Refractive Index | 1.627 |
Show More |
RUO – Research Use Only