missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, HPLC Grade, 99.8+%, Thermo Scientific Chemicals

Catalog Number p-7050609
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S
This item is not returnable. View return policy
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S

Carbon disulfide is used in spectrophotometry, high performance liquid chromatography and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -112°C
Assay Percent Range >99.8%
UN Number UN1131
Merck Index 14,1811
Solubility Information Miscible with alcohol,ether,benzene,oil,chloroform and carbon tetrachloride.
Formula Weight 76.14
Percent Purity ≥99.8%
Grade HPLC
Density 1.2632 g/mL
Boiling Point 46°C
Quantity 1 L
Flash Point -30°C (-22°F)
Odor Unpleasant
Physical Form Liquid
Refractive Index 1.627
Chemical Name or Material Carbon disulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'