missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen

Catalog Number 15672860
Click to view available options
Quantity:
100 mL
1 L
4 x 2 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S
This item is not returnable. View return policy
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY
Melting Point -112°C to -111°C (literature)
Absorbance max. 0.50 at 388 nm, max. 1.0 at 386 nm, max. 0.01 at 750 nm, max. 0.01 500 nm, max. 0.05 at 410 nm, max. 0.10 at 403 nm, max. 0.25 at 394 nm
CAS Max % 100.0000%
Linear Formula CS2
UN Number UN1131
Beilstein 1098293
Formula Weight 76.14
Percent Purity >99.9%
Grade HPLC
Density 1.266 g/mL at 25 °C(lit.)
Boiling Point 46°C
Quantity 100 mL
Flash Point −30°C
Packaging Glass Bottle
Chemical Name or Material Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'