missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon disulfide, ACS Reagent, ≥99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -76°C |
Color | Colorless |
UN Number | UN1131 |
Beilstein | 1098293 |
Solubility Information | 2.9g/L in water at 20°C |
Formula Weight | 76.14g/mol |
Percent Purity | ≥99.9% |
Grade | ACS Reagent |
Density | 1.266g/mL at 25°C |
Boiling Point | 46°C (1013 hPa) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'