Learn More
Activated charcoal, NORIT™ CN 1, Acros Organics™
Brand: Acros Organics 404042500
Code : 29
Additional Details : CAS Number : 7440-44-0 Weight : 0.30000kg Transport : UN number : 1362 Chem class : 1 Pack group : III
Quantity | 250g |
---|---|
Packaging | Glass bottle |
Description
Filter aidChemical Identifiers
Activated charcoal, NORIT™ CN 1 | |
7440-44-0 | |
12 | |
Carbon, activated |
NORIT CN 1 | |
C | |
MFCD00133992 |
Specifications
Activated charcoal, NORIT™ CN 1 | |
3652.0°C | |
5.5 to 8 | |
7440-44-0 | |
200ppm max. | |
150ppm max. | |
C | |
MFCD00133992 | |
15, 1807 | |
Carbon, activated | |
trace analysis by acid extraction |
Black | |
250g | |
NORIT CN 1 | |
methylene blue adsorption (g/100g):> 25 | |
Powder | |
15% max. | |
12 | |
Glass bottle | |
3.5% max. | |
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents |