missing translation for 'onlineSavingsMsg'
Learn More

(Carbethoxymethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

Catalog Number 10496900
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1099-45-2
C22H21O2P
348.38
MFCD00009183
IIHPVYJPDKJYOU-UHFFFAOYSA-N
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
70670
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1099-45-2
C22H21O2P
348.38
MFCD00009183
IIHPVYJPDKJYOU-UHFFFAOYSA-N
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
70670
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 124.0°C to 129.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCO2C2H5
Quantity 25 g
Fieser 01,112; 02,60; 10,78
Solubility Information Solubility in water: insoluble
Formula Weight 348.38
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material (Carbethoxymethylene)triphenylphosphorane
Show More Show Less
compliance-icons
Product Identifier
  • (Carbethoxymethylene)triphenylphosphorane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'